Learn More
CPC Scientific DNP-Gly-Pro-Leu-Gly-Met-Arg-Gly-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MMPS015A
Encompass
Highly sensitive and selective substrate for collagenase 3 (MMP-13) with a kcat/Km value of 4.22 x 106 M-1s-1 (PH= 7.5, T = 37°C)ONE-LETTER SEQUENCE: Dnp-GPLGMRGL-NH2MOLECULAR FORMULA: C40H64N14O12S1MOLECULAR WEIGHT:965.11STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1872435-02-3], SYNONYMS: Dnp-Gly-Pro-Leu-Gly-Met-Arg-Gly-Leu-NH2, Dnp-GPLGMRGL-NH2, Matrix Metalloproteinase-13 SubstrateRESEARCH AREA: CancerREFERENCES: S.J. Deng, et al., J. Biol. Chem., 275, 31422-31427 (2000)
Catalog No. 50-210-9777