Learn More
CPC Scientific MCA-Pro-Cha-Gly-Nva-His-Ala-Dap(DNP)-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109780
Encompass
Specific fluorogenic substrate for collagenase-3 (MMP) with a kcat/Km value of 1.09 x 106 M-1s-1 (pH = 7.5, T = 25 °C, substrate concentrations: 0.7 and 1.4 µM (S<
Catalog No. 50-210-9780 Supplier CPC Scientific Supplier No. MMPS016B
Specific fluorogenic substrate for collagenase-3 (MMP) with a kcat/Km value of 1.09 x 106 M-1s-1 (pH = 7.5, T = 25 °C, substrate concentrations: 0.7 and 1.4 µM (S<