Learn More
Enzo Life Sciences Matrix Metalloproteinase-14 (MMP-14) fluorometric drug discovery kit (1x96wells)
SDP
Catalog No. 502010277
Encompass_Preferred
A QUANTIZYME Assay System. The MMP-14 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay sytem designed to screen for inhibitors of matrix metalloproteinase-14 (MMP-14, membrane-type matrix metalloproteinase 1, MT1-MMP) using a quenched fluorogenic peptide: OMNIMMP fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. Also included are active enzyme, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet. Alternative name: Membrane type matrix metalloproteinase 1, MT-1 MMP. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0277 Supplier Enzo Life Sciences Supplier No. BMLAK4170001
A QUANTIZYME Assay System. The MMP-14 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay sytem designed to screen for inhibitors of matrix metalloproteinase-14 (MMP-14, membrane-type matrix metalloproteinase 1, MT1-MMP) using a quenched fluorogenic peptide: OMNIMMP fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. Also included are active enzyme, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet. Alternative name: Membrane type matrix metalloproteinase 1, MT-1 MMP. Applications: Fluorescent detection, HTS Activity assay.