Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MMP-19 protein, human | >85.0% | 10 UG
SDP

Catalog No. 5000291949
Encompass_Preferred

Recombinant human MMP-19 protein, tag-free, supplied as a sterile filtered solution for research use. The protein is expressed in E. coli, provided in microgram quantities, and formulated for biochemical and molecular biology applications; storage at -80°C is recommended and repeated freeze-thaw cycles should be avoided.

  • Tag-free recombinant human protein expressed in E. coli.
  • Supplied as a 0.22 μm filtered solution formulated in 0.1% TFA and 30% acetonitrile.
  • Available in microgram quantities suitable for biochemical assays.
  • Purity greater than 85% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~18 kDa (reducing conditions).
  • Includes datasheet and safety data sheet for handling and storage recommendations.
  • Store at -80°C and avoid repeated freeze-thaw cycles; stable at -20°C for short-term use.

Catalog No. 50-002-91949 Supplier Medchemexpress LLC Supplier No. HYP7592910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human MMP-19 protein, tag-free, supplied as a sterile filtered solution for research use. The protein is expressed in E. coli, provided in microgram quantities, and formulated for biochemical and molecular biology applications; storage at -80°C is recommended and repeated freeze-thaw cycles should be avoided.

  • Tag-free recombinant human protein expressed in E. coli.
  • Supplied as a 0.22 μm filtered solution formulated in 0.1% TFA and 30% acetonitrile.
  • Available in microgram quantities suitable for biochemical assays.
  • Purity greater than 85% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~18 kDa (reducing conditions).
  • Includes datasheet and safety data sheet for handling and storage recommendations.
  • Store at -80°C and avoid repeated freeze-thaw cycles; stable at -20°C for short-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.