Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MMP-19 protein, human | >85.0% | 100 UG
SDP

Catalog No. 5000290967
Encompass_Preferred

Recombinant human MMP-19 protein supplied as a filtered solution for research use. The protein is tag free, expressed in E. coli, and provided at 100 μg in a formulation designed for stability in storage and shipment on dry ice.

  • Provided as a 0.22 μm filtered solution (0.1% TFA, 30% CAN)
  • Tag free and expressed in E. coli
  • Purity greater than 85% by reducing SDS-PAGE
  • Molecular weight ~18 kDa (reducing conditions)
  • Endotoxin <1 EU/μg (LAL method)
  • Store at -80°C; shipped on dry ice
  • Enzyme activity testing in progress and not guaranteed

Catalog No. 50-002-90967 Supplier Medchemexpress LLC Supplier No. HYP75929100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human MMP-19 protein supplied as a filtered solution for research use. The protein is tag free, expressed in E. coli, and provided at 100 μg in a formulation designed for stability in storage and shipment on dry ice.

  • Provided as a 0.22 μm filtered solution (0.1% TFA, 30% CAN)
  • Tag free and expressed in E. coli
  • Purity greater than 85% by reducing SDS-PAGE
  • Molecular weight ~18 kDa (reducing conditions)
  • Endotoxin <1 EU/μg (LAL method)
  • Store at -80°C; shipped on dry ice
  • Enzyme activity testing in progress and not guaranteed

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.