Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MMP-19 protein, human | >85.0% | 5 UG
SDP

Catalog No. 5000291967
Encompass_Preferred

Recombinant human MMP-19 protein supplied as a tag-free, purified solution for research use. The product corresponds to amino acids L101-G256 of the human sequence (accession Q99542-1), with an approximate molecular weight of 18 kDa, and is provided in a filtered solvent suitable for biochemical assays and antibody development. Store frozen and avoid repeated freeze-thaw.

  • Tag free protein corresponding to L101-G256 of human MMP-19.
  • Expression system: Escherichia coli.
  • Purity >85% by reducing SDS-PAGE.
  • Approximate molecular weight 18 kDa.
  • Formulation: 0.22 μm filtered solution in 0.1% TFA, 30% ACN.
  • Recommended storage at -80 °C; avoid repeated freeze-thaw.
  • For research use only; not for clinical applications.

Catalog No. 50-002-91967 Supplier Medchemexpress LLC Supplier No. HYP759295UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human MMP-19 protein supplied as a tag-free, purified solution for research use. The product corresponds to amino acids L101-G256 of the human sequence (accession Q99542-1), with an approximate molecular weight of 18 kDa, and is provided in a filtered solvent suitable for biochemical assays and antibody development. Store frozen and avoid repeated freeze-thaw.

  • Tag free protein corresponding to L101-G256 of human MMP-19.
  • Expression system: Escherichia coli.
  • Purity >85% by reducing SDS-PAGE.
  • Approximate molecular weight 18 kDa.
  • Formulation: 0.22 μm filtered solution in 0.1% TFA, 30% ACN.
  • Recommended storage at -80 °C; avoid repeated freeze-thaw.
  • For research use only; not for clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.