Learn More
CPC Scientific MCA-Pro-Leu-Gly-Leu-Dap(DNP)-Ala-Arg-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109763
Encompass
SEQUENCE: MCA-Pro-Leu-Gly-Leu-Dap(DNP)-Ala-Arg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-PLGL-Dap(Dnp)-AR-NH2MOLECULAR FORMULA: C49H68N14O15MOLECULAR WEIGHT: 1093.17STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [140430-53-1]SYNONYMS: 7-Methoxycoumarin-4-Acetyl-Pro-Leu-Gly-Leu-Dnp-(2,3-Diaminopropionyl)-Ala-Arg, amideRESEARCH AREA: CancerREFERENCES:L.Zhu et al., Arch. Oral Biol., 53, 785 (2008)H.Oneda et al., Biosci. Biotechnol. Biochem., 68, 1811 (2004)Z.S.Galis et al., FASEB J., 9, 974 (1995)C.G.Knight et al., FEBS Lett., 296, 263 (1992) SKU(s): MMPS-009A, MMPS-009B
Catalog No. 50-210-9763 Supplier CPC Scientific Supplier No. MMPS009A
Add to Cart
Add to Cart
SEQUENCE: MCA-Pro-Leu-Gly-Leu-Dap(DNP)-Ala-Arg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-PLGL-Dap(Dnp)-AR-NH2MOLECULAR FORMULA: C49H68N14O15MOLECULAR WEIGHT: 1093.17STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [140430-53-1]SYNONYMS: 7-Methoxycoumarin-4-Acetyl-Pro-Leu-Gly-Leu-Dnp-(2,3-Diaminopropionyl)-Ala-Arg, amideRESEARCH AREA: CancerREFERENCES:L.Zhu et al., Arch. Oral Biol., 53, 785 (2008)H.Oneda et al., Biosci. Biotechnol. Biochem., 68, 1811 (2004)Z.S.Galis et al., FASEB J., 9, 974 (1995)C.G.Knight et al., FEBS Lett., 296, 263 (1992) SKU(s): MMPS-009A, MMPS-009B