Learn More
CPC Scientific MCA-Pro-Leu-Gly-Leu-Dap(DNP)-Ala-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MMPS009B
Encompass
SEQUENCE: MCA-Pro-Leu-Gly-Leu-Dap(DNP)-Ala-Arg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-PLGL-Dap(Dnp)-AR-NH2MOLECULAR FORMULA: C49H68N14O15MOLECULAR WEIGHT: 1093.17STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [140430-53-1]SYNONYMS: 7-Methoxycoumarin-4-Acetyl-Pro-Leu-Gly-Leu-Dnp-(2,3-Diaminopropionyl)-Ala-Arg, amideRESEARCH AREA: CancerREFERENCES:L.Zhu et al., Arch. Oral Biol., 53, 785 (2008)H.Oneda et al., Biosci. Biotechnol. Biochem., 68, 1811 (2004)Z.S.Galis et al., FASEB J., 9, 974 (1995)C.G.Knight et al., FEBS Lett., 296, 263 (1992) SKU(s): MMPS-009A, MMPS-009B
Catalog No. 50-210-9764