Learn More
CPC Scientific MCA-Pro-Leu-Gly-Leu-Dap-Ala-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109760
Encompass
ONE-LETTER SEQUENCE: Mca-PLGL-Dap-AR-NH2MOLECULAR FORMULA: C43H66N12O11MOLECULAR WEIGHT:927.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: C.G. Knight, et al. FEBS Lett. 296, 263 (1992)
Catalog No. 50-210-9760 Supplier CPC Scientific Supplier No. MMPS007B
ONE-LETTER SEQUENCE: Mca-PLGL-Dap-AR-NH2MOLECULAR FORMULA: C43H66N12O11MOLECULAR WEIGHT:927.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: C.G. Knight, et al. FEBS Lett. 296, 263 (1992)