Learn More
CPC Scientific H-Cys-Thr-Thr-His-Trp-Gly-Phe-Thr-Leu-Cys-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109765
Encompass
Inhibitor of gelatinases but not other MMP family members. When combined with tumor targeting, anti-invasive properties, it became a potential lead compound for the development of peptide-mimetic anticancer agents. ONE-LETTER SEQUENCE: CTTHWGFTLC (Cys1 and 10 bridge)MOLECULAR FORMULA: C52H71N13O14S2MOLECULAR WEIGHT:1166.33STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [244082-19-7], RESEARCH AREA: CancerREFERENCES: E.Koivunen et al, Nature Biotechnol., 17, 768 (1999)
Catalog No. 50-210-9765 Supplier CPC Scientific Supplier No. MMPS010A
Inhibitor of gelatinases but not other MMP family members. When combined with tumor targeting, anti-invasive properties, it became a potential lead compound for the development of peptide-mimetic anticancer agents. ONE-LETTER SEQUENCE: CTTHWGFTLC (Cys1 and 10 bridge)MOLECULAR FORMULA: C52H71N13O14S2MOLECULAR WEIGHT:1166.33STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [244082-19-7], RESEARCH AREA: CancerREFERENCES: E.Koivunen et al, Nature Biotechnol., 17, 768 (1999)