Learn More
CPC Scientific DNP-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MMPS011A
Encompass
SEQUENCE: DNP-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: DNP-PLGMWSRMOLECULAR FORMULA: C44H61N13O13S1MOLECULAR WEIGHT: 1012.1STORAGE CONDITIONS:-20 5 CCAS REGISTRY NUMBER: [135662-07-6]SYNONYMS: 2,4-Dinitrophenyl-Pro-Leu-Gly-Met-Trp-Ser-ArgRESEARCH AREA: CancerCATEGORY: MMP Substrates REFERENCES:S. Netzel-Arnett, et al., Anal. Biochem. 195, 86 (1991)SKU(s): MMPS-011A, MMPS-011B, MMPS-011C, MMPS-011D
Catalog No. 50-210-9767