Learn More
Medchemexpress LLC Matrix metalloproteinase-2 (rat), recombinant (His tag) | 90.0% | 50UG
SDP
Catalog No. 5000176467
Encompass_Preferred
Recombinant rat matrix metalloproteinase-2 (MMP-2) expressed in Pichia pastoris with a His tag, supplied as a 50 μg vial for research use. Suitable for in vitro biochemical assays and cell-based studies of extracellular matrix remodeling.
  • Supplied as 50 μg vial.
  • Recombinant protein expressed in Pichia pastoris.
  • His tag for purification.
  • Reported purity 90% (distributor listing).
  • Approximate molecular weight 64.1 kDa (listed value).
  • Intended for in vitro biochemical and cell biology applications.
Catalog No. 50-001-76467 Supplier Medchemexpress LLC Supplier No. HYP70057350UG
Recombinant rat matrix metalloproteinase-2 (MMP-2) expressed in Pichia pastoris with a His tag, supplied as a 50 μg vial for research use. Suitable for in vitro biochemical assays and cell-based studies of extracellular matrix remodeling.
  • Supplied as 50 μg vial.
  • Recombinant protein expressed in Pichia pastoris.
  • His tag for purification.
  • Reported purity 90% (distributor listing).
  • Approximate molecular weight 64.1 kDa (listed value).
  • Intended for in vitro biochemical and cell biology applications.