Learn More
Enzo Life Sciences MMP-20 (catalytic domain) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5400010
Encompass_Preferred
Alternative name: Matrix metalloproteinase 20, Enamelysin. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, 5mM CaCl2, 300mM NaCl, 20µM ZnCl2, 0.5% Brij-35, and 30% glycerol. Source: Produced in E. coli. Active recombinant matrix metalloproteinase-20 (MMP-20, enamelysin) cloned from human cDNA. The enzyme consists of residues Tyr108-Phe276 (NM_004771), which comprises the catalytic domain of human MMP-20, with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-20 which lacks the C-terminal hemopexin domain. Long Term Storage: -80°C.
Catalog No. 50-201-2076