Learn More
Enzo Life Sciences MMP-3 inhibitor (5mg). CAS: 158841-76-0
SDP
Catalog No. 502008363
Encompass_Preferred
Inhibitor of matrix metalloproteinase 3 (MMP-3) (IC50=5µM). Alternative name: Ac-RCGVPD-NH2, Stromelysin 1 inhibitor, Matrix metalloproteinase 3 inhibitor. Formula: C27H46N10O9S. MW: 686.8. Purity: ≥95% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8363 Supplier Enzo Life Sciences Supplier No. ALX260165M005
Inhibitor of matrix metalloproteinase 3 (MMP-3) (IC50=5µM). Alternative name: Ac-RCGVPD-NH2, Stromelysin 1 inhibitor, Matrix metalloproteinase 3 inhibitor. Formula: C27H46N10O9S. MW: 686.8. Purity: ≥95% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water. Long Term Storage: -20°C.