Learn More
Enzo Life Sciences MMP-7 (catalytic domain) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE1810010
Encompass_Preferred
Alternative name: Matrix metalloproteinase 7, Matrilysin, Pump. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, 5mM CaCl2, 300mM NaCl, 20µM ZnCl2, 0.5% Brij-35, and 30% glycerol. Source: Produced in E. coli. Active Matrix Metalloproteinase-7 (MMP-7, matrilysin, pump) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-7 (Tyr95-Lys267, NM_002423) with a C-terminal purification tag. Long Term Storage: -80°C.
Catalog No. 50-201-1953