Learn More
CPC Scientific DNP-Arg-Pro-Leu-Ala-Leu-Trp-Arg-Ser-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109774
Encompass
Best fluorogenic substrate developed for matrilysin (also called punctuated metalloproteinase 1, PUMP-1 or MMP-7) thus far. Besides good kinetic parameters (kcat/km= 1.9 x 105 M-1s-1), it presents good solubility in assay buffer.SEQUENCE: DNP-Arg-Pro-Leu-Ala-Leu-Trp-Arg-Ser-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: DNP-RPLALWRSMOLECULAR FORMULA: C52H77N17O14MOLECULAR WEIGHT: 1164.30STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [172666-82-9]SYNONYMS: MMP-7 Fluorogenic SubstrateRESEARCH AREA: CancerREFERENCES:A.R. Welch, et al., Arch. Biochem. Biophys. 324, 59 (1995)SKU(s): MMPS-013A, MMPS-013B
Catalog No. 50-210-9774 Supplier CPC Scientific Supplier No. MMPS013B
Best fluorogenic substrate developed for matrilysin (also called punctuated metalloproteinase 1, PUMP-1 or MMP-7) thus far. Besides good kinetic parameters (kcat/km= 1.9 x 105 M-1s-1), it presents good solubility in assay buffer.SEQUENCE: DNP-Arg-Pro-Leu-Ala-Leu-Trp-Arg-Ser-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: DNP-RPLALWRSMOLECULAR FORMULA: C52H77N17O14MOLECULAR WEIGHT: 1164.30STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [172666-82-9]SYNONYMS: MMP-7 Fluorogenic SubstrateRESEARCH AREA: CancerREFERENCES:A.R. Welch, et al., Arch. Biochem. Biophys. 324, 59 (1995)SKU(s): MMPS-013A, MMPS-013B