Learn More
Enzo Life Sciences Matrix Metalloproteinase-8 (MMP-8) fluorometric drug discovery kit (1x96wells)
SDP
Catalog No. 502010275
Encompass_Preferred
A QUANTIZYME Assay System. The MMP-8 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) using a quenched fluorogenic peptide: OMNIMMP fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. Also included are active enzyme, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet. Alternative name: Neutrophil collagenase, Collagenase 2. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0275 Supplier Enzo Life Sciences Supplier No. BMLAK4150001
A QUANTIZYME Assay System. The MMP-8 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) using a quenched fluorogenic peptide: OMNIMMP fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. Also included are active enzyme, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet. Alternative name: Neutrophil collagenase, Collagenase 2. Applications: Fluorescent detection, HTS Activity assay.