Learn More
Medchemexpress LLC MMP-8 Human HEK293 10ug
SDP
Catalog No. 5000181488
Encompass_Preferred
MMP-8 also known as matrix metalloproteinase-8 or collagenase-2 is recognized for its enzymatic abilities particularly its ability to break down fibrillar types I II and III collagen This enzymatic activity suggests that MMP-8 plays an important role in the remodeling and turnover of collagen-rich tissues such as connective tissue cartilage and bone MMP-8 Protein Human (HEK293 His) is the recombinant human-derived MMP-8 protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-81488 Supplier Medchemexpress LLC Supplier No. HYP7250710UG
MMP-8 also known as matrix metalloproteinase-8 or collagenase-2 is recognized for its enzymatic abilities particularly its ability to break down fibrillar types I II and III collagen This enzymatic activity suggests that MMP-8 plays an important role in the remodeling and turnover of collagen-rich tissues such as connective tissue cartilage and bone MMP-8 Protein Human (HEK293 His) is the recombinant human-derived MMP-8 protein expressed by HEK293 with C-10 His labeled tag