Learn More
CPC Scientific DNP-Pro-Leu-Ala-Tyr-Trp-Ala-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109775
Encompass
Fluorogenic substrate for human neutrophil collagenase (HNC; MMP-8).SEQUENCE: DNP-Pro-Leu-Ala-Tyr-Trp-Ala-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-PLAYWARMOLECULAR FORMULA: C49H63N13O13MOLECULAR WEIGHT: 1042.12STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [135662-05-4]SYNONYMS: RESEARCH AREA: CancerREFERENCES:S. Netzel-Arnett, et al., Anal. Biochem. 195, 86 (1991)SKU(s): MMPS-014A, MMPS-014B
Catalog No. 50-210-9775 Supplier CPC Scientific Supplier No. MMPS014A
Fluorogenic substrate for human neutrophil collagenase (HNC; MMP-8).SEQUENCE: DNP-Pro-Leu-Ala-Tyr-Trp-Ala-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-PLAYWARMOLECULAR FORMULA: C49H63N13O13MOLECULAR WEIGHT: 1042.12STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [135662-05-4]SYNONYMS: RESEARCH AREA: CancerREFERENCES:S. Netzel-Arnett, et al., Anal. Biochem. 195, 86 (1991)SKU(s): MMPS-014A, MMPS-014B