Learn More
Enzo Life Sciences Matrix Metalloproteinase-9 (MMP-9) fluorometric drug discovery kit (1x96wells)
SDP
Catalog No. NC1422757
Encompass_Preferred
A QUANTIZYME Assay System. The MMP-9 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-9 (MMP-9, gelatinase B) using a quenched fluorogenic peptide: OMNIMMP fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-9, a potential therapeutic target. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH)3, and a detailed instruction booklet. Alternative Name: Gelatinase B, 92kDa Type IV collagenase. Applications: Fluorescent detection, HTS, Activity assay. Contents: MMP-9 enzyme (human) (recombinant), substrate, calibration standard, inhibitor, assay buffer, 96-well white microplate.
Catalog No. NC1422757 Supplier Enzo Life Sciences Supplier No. BMLAK4110001
A QUANTIZYME Assay System. The MMP-9 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-9 (MMP-9, gelatinase B) using a quenched fluorogenic peptide: OMNIMMP fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-9, a potential therapeutic target. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH)3, and a detailed instruction booklet. Alternative Name: Gelatinase B, 92kDa Type IV collagenase. Applications: Fluorescent detection, HTS, Activity assay. Contents: MMP-9 enzyme (human) (recombinant), substrate, calibration standard, inhibitor, assay buffer, 96-well white microplate.