Learn More
BioVendor, LLC Mmp-9 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL002R
Encompass
Increased matrix metalloprotease 9 (MMP9), also called Gelatinase B, after myocardial infarction (MI) exacerbates ischemia-induced chronic heart failure (CHF). It is also upregulated in the diabetic heart, and ablation of MMP9 decreases infarct size in the non-diabetic myocardial infarction heart.3 There is also strong evidence that the increased levels of MMP-9 are associated with many inflammatory conditions, such as coronary artery disease, COPD, arthritis, and metabolic syndrome. It was also proved that MMP-9 is associated with progression of atherosclerosis and higher risk of cardiovascular events. It was also suggested that elevated levels of MMP-9 (combined with MMP-2) are associated with highly tumorigenic cancers. One kit (96 wells). Type: sandwich ELISA. Sample type: serum, plasma. Sample req: 5 ¿l/well. Cal range: 0.625-20ng/ml. LoD: 0.16ng/ml. Other names: Matrix Metalloproteinase-9, MMP9.
Catalog No. 50-240-6365