Learn More
Medchemexpress LLC MMP-9 Human HEK293 100ug
SDP
Catalog No. 5000180333
Encompass_Preferred
MMP-9 is an important member of the MMP protein family and regulates the extracellular matrix during physiological processes such as development and tissue remodeling It involves arthritis and metastasis MMP-9 Protein Human (HEK293) is the recombinant human-derived MMP-9 protein expressed by HEK293 with tag free
Catalog No. 50-001-80333 Supplier Medchemexpress LLC Supplier No. HYP73300100UG
MMP-9 is an important member of the MMP protein family and regulates the extracellular matrix during physiological processes such as development and tissue remodeling It involves arthritis and metastasis MMP-9 Protein Human (HEK293) is the recombinant human-derived MMP-9 protein expressed by HEK293 with tag free