Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MMP-9 protein, human (P. pastoris, His) | >90.0% | 50UG
SDP

Catalog No. 5000176621
Encompass_Preferred

Recombinant human matrix metalloproteinase-9 (MMP-9) expressed in Pichia pastoris and purified with a His tag. Supplied as a lyophilized powder for research use, MMP-9 is involved in localized extracellular matrix breakdown and leukocyte migration. Enzymatic activity testing is in progress and activity is not guaranteed by the manufacturer.

  • Expression system: P. pastoris.
  • His-tagged for affinity purification.
  • Molecular weight: 69 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg (LAL).
  • Formulation: lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Storage: store at -20°C; stable up to 2 years frozen.
  • Reconstitution: avoid concentrations below 100 μg/mL in ddH2O; aliquot for long-term storage.

Catalog No. 50-001-76621 Supplier Medchemexpress LLC Supplier No. HYP70057250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human matrix metalloproteinase-9 (MMP-9) expressed in Pichia pastoris and purified with a His tag. Supplied as a lyophilized powder for research use, MMP-9 is involved in localized extracellular matrix breakdown and leukocyte migration. Enzymatic activity testing is in progress and activity is not guaranteed by the manufacturer.

  • Expression system: P. pastoris.
  • His-tagged for affinity purification.
  • Molecular weight: 69 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg (LAL).
  • Formulation: lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Storage: store at -20°C; stable up to 2 years frozen.
  • Reconstitution: avoid concentrations below 100 μg/mL in ddH2O; aliquot for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.