Learn More
Medchemexpress LLC HY-135232 5mg Medchemexpress, MMP-9-IN-1 CAS:502887-71-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1352325MG
Encompass_Preferred
Medchemexpress, HY-135232 5mg MMP-9-IN-1 CAS:502887-71-0 MMP-9-IN-1 is a specific matrix metalloproteinase-9 (MMP-9) inhibitor, which selectively target the hemopexin (PEX) domain of MMP-9, but not other MMPs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9868