Learn More
Enzo Life Sciences MMP-​9 inhibitor I (1mg). CAS: 206549-55-5
SDP
Catalog No. 502013426
Encompass_Preferred
Alternative name: 2-(N-benzyl-4-methoxyphenylsulfonamido)-5-((diethylamino)methyl)-N-hydroxy-3-methylbenzamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3426 Supplier Enzo Life Sciences Supplier No. ENZCHM1330001
Alternative name: 2-(N-benzyl-4-methoxyphenylsulfonamido)-5-((diethylamino)methyl)-N-hydroxy-3-methylbenzamide. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.