Learn More
Sigma Aldrich Fine Chemicals Biosciences MMP 9 pre activated human
Catalog No. 502905425
Encompass
Recombinant human pre-activated Matrix Metalloproteinase-9 (MMP-9) is expressed in human HEK 293 cells as a glycoprotein. The DTT-reduced protein migrates as a ~67 kDa polypeptide on SDS-PAGE. This protein is manufactured in human cells with no serum. The human cells expression system allows human-like glycosylation and folding and often supports higher specific activity of the protein. The protein is produced with no artificial tags.
Catalog No. 50-290-5425 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE007850UG
Recombinant human pre-activated Matrix Metalloproteinase-9 (MMP-9) is expressed in human HEK 293 cells as a glycoprotein. The DTT-reduced protein migrates as a ~67 kDa polypeptide on SDS-PAGE. This protein is manufactured in human cells with no serum. The human cells expression system allows human-like glycosylation and folding and often supports higher specific activity of the protein. The protein is produced with no artificial tags.