Learn More
Enzo Life Sciences MMP-Inhibitor profiling kit, fluorometric (1x96wells)
SDP
Catalog No. 502010246
Encompass_Preferred
A QUANTIZYME Assay System Complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes. The assays are performed in a convenient 96-well microplate format using a sensitive fluorogenic substrate . Included are active MMP enzymes, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0246 Supplier Enzo Life Sciences Supplier No. BMLAK0160001
A QUANTIZYME Assay System Complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes. The assays are performed in a convenient 96-well microplate format using a sensitive fluorogenic substrate . Included are active MMP enzymes, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet. Applications: Fluorescent detection, HTS Activity assay.