Learn More
CPC Scientific H-Abz-Gly-Pro-DLeu-DAla-NH-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109746
Encompass
Inhibitor of human matrix metalloproteinases that maintains its activity even after prolonging incubation with pronase or human granulocyte elastase. It also inhibits interstitial and granulocyte collagenases, granulocyte gelatinase and skin fibroblast stromelysin with IC50 of 1 x 10-6 M, 3 x 10-5 M and 1.5 x 10-4 M, respectively.SEQUENCE: H-Abz-Gly-Pro-DLeu-DAla-NH-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Abz-GP-DLeu-DAla-NH-OHMOLECULAR FORMULA: C23H35N6O6MOLECULAR WEIGHT: 490.55STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [124168-73-6]SYNONYMS: 4-Abz-Gly-Pro-D-Leu-D-Ala-NHOHRESEARCH AREA: CancerREFERENCES:S. Odake, et al., Biochem. Biophys. Res. Commun. 199, 1442 (1994)SKU(s): MMPS-001A, MMPS-001B
Catalog No. 50-210-9746 Supplier CPC Scientific Supplier No. MMPS001B
Inhibitor of human matrix metalloproteinases that maintains its activity even after prolonging incubation with pronase or human granulocyte elastase. It also inhibits interstitial and granulocyte collagenases, granulocyte gelatinase and skin fibroblast stromelysin with IC50 of 1 x 10-6 M, 3 x 10-5 M and 1.5 x 10-4 M, respectively.SEQUENCE: H-Abz-Gly-Pro-DLeu-DAla-NH-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Abz-GP-DLeu-DAla-NH-OHMOLECULAR FORMULA: C23H35N6O6MOLECULAR WEIGHT: 490.55STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [124168-73-6]SYNONYMS: 4-Abz-Gly-Pro-D-Leu-D-Ala-NHOHRESEARCH AREA: CancerREFERENCES:S. Odake, et al., Biochem. Biophys. Res. Commun. 199, 1442 (1994)SKU(s): MMPS-001A, MMPS-001B