Learn More
Enzo Life Sciences Matrix metalloproteinase (MMP) multipack-1 (1 Kit)
SDP
Catalog No. NC1165118
Encompass_Preferred
The MMP multipack-1 contains 10µg each of five highly active recombinant MMP catalytic domains: MMP-1, MMP-2, MMP-8, MMP-9, and MMP-13. Contents: Contains 10µg of each enzyme, provided in a screw-cap microfuge tube. MMP-1 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE180), MMP-2 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE237), MMP-8 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE255), MMP-9 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE360), MMP-13 (catalytic domain) (human), (recombinant) (Prod. N. BML-S24). Application: FUNC, HTS. Long Term Storage: -80°C
Catalog No. NC1165118 Supplier Enzo Life Sciences Supplier No. BMLAK0130001
The MMP multipack-1 contains 10µg each of five highly active recombinant MMP catalytic domains: MMP-1, MMP-2, MMP-8, MMP-9, and MMP-13. Contents: Contains 10µg of each enzyme, provided in a screw-cap microfuge tube. MMP-1 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE180), MMP-2 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE237), MMP-8 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE255), MMP-9 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE360), MMP-13 (catalytic domain) (human), (recombinant) (Prod. N. BML-S24). Application: FUNC, HTS. Long Term Storage: -80°C