Learn More
Enzo Life Sciences Matrix metalloproteinase (MMP) multipack-2 (1 Kit)
SDP
Catalog No. NC1635961
Encompass_Preferred
The MMP multipack-2 contains 10µg each of five highly active recombinant MMP catalytic domains: MMP-3, MMP-7, MMP-10, MMP-11, and MMP-12. Contents: Contains 10µg of each enzyme, provided in a screw-cap microfuge tube. MMP-3 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE109), MMP-7 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE181), MMP-10 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE392), MMP-11  (catalytic domain) (human), (recombinant) (Prod. No. BML-SE282), MMP-12 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE138). Application: FUNC, HTS. Long Term Storage: -80°C
Catalog No. NC1635961 Supplier Enzo Life Sciences Supplier No. BMLAK0140001
The MMP multipack-2 contains 10µg each of five highly active recombinant MMP catalytic domains: MMP-3, MMP-7, MMP-10, MMP-11, and MMP-12. Contents: Contains 10µg of each enzyme, provided in a screw-cap microfuge tube. MMP-3 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE109), MMP-7 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE181), MMP-10 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE392), MMP-11  (catalytic domain) (human), (recombinant) (Prod. No. BML-SE282), MMP-12 (catalytic domain) (human), (recombinant) (Prod. No. BML-SE138). Application: FUNC, HTS. Long Term Storage: -80°C