Learn More
Echelon Biosciences Research Labs MMP Substrate - 1 mg, CAS#: 720710-69-0
SDP
Supplier:  Echelon Biosciences Research Labs 868411MG
Encompass_Preferred
MMP Substrate - 1mg , Mca-Lys-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2 Category/Application: Enzyme Substrates This item is non returnable
Catalog No. 50-190-8968