Learn More
Enzo Life Sciences MMP-substrate (fluorogenic) (1mg)
SDP
Catalog No. 502011241
Encompass_Preferred
Fluorogenic substrate for several matrix metalloproteinases (MMPs). It is cleaved by MMP-1, MMP-3, MMP-7, MMP-8, MMP-9, MMP-11, MMP-12, MMP-13, and ADAM9/MDC9, but only slightly by MMP-2 and not at all by ADAM17/TACE. Ex/Em= 340nm/440nm, although 365/450 is also appropriate. Highly quenched, with strong fluorescence by Nma at 440nm once cleavage separates it from the aromatic Dnp moiety. Compatible with most fluorometers. Alternative name: Matrix metalloproteinase substrate, Dnp-Pro-beta-cyclohexyl-Ala-Gly-Cys(Me)-His-Ala-Lys(Nma)-NH2. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (at least 20mM). Long Term Storage: -20°C.
Catalog No. 50-201-1241 Supplier Enzo Life Sciences Supplier No. BMLP1280001
Fluorogenic substrate for several matrix metalloproteinases (MMPs). It is cleaved by MMP-1, MMP-3, MMP-7, MMP-8, MMP-9, MMP-11, MMP-12, MMP-13, and ADAM9/MDC9, but only slightly by MMP-2 and not at all by ADAM17/TACE. Ex/Em= 340nm/440nm, although 365/450 is also appropriate. Highly quenched, with strong fluorescence by Nma at 440nm once cleavage separates it from the aromatic Dnp moiety. Compatible with most fluorometers. Alternative name: Matrix metalloproteinase substrate, Dnp-Pro-beta-cyclohexyl-Ala-Gly-Cys(Me)-His-Ala-Lys(Nma)-NH2. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (at least 20mM). Long Term Storage: -20°C.