Learn More
CPC Scientific DNP-Pro-Leu-Gly-Leu-Trp-Ala-DArg-NH2 (trifluoroacetate salt) 25MG
SDP
Catalog No. 502109790
Encompass
Efficient substrate for intestitial/vertebrate collagenase (MMP-1), stromelysin (MMP-3), gelatinases (MMP-2 and MMP-9) and the punctuated metalloproteinase (MMP-7).SEQUENCE: DNP-Pro-Leu-Gly-Leu-Trp-Ala-DArg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-PLGLWA-DArg-NH2MOLECULAR FORMULA: C45H64N14O11MOLECULAR WEIGHT: 977.09STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [121282-17-5]SYNONYMS: Dnp-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg-NHRESEARCH AREA: CancerREFERENCES:D.M. Bickett, et al., Anal. Biochem. 212, 58 (1993)C.G. Knight, et al., FEBS Lett. 296, 263 (1992)K. Darlak, et al., J. Biol. Chem. 265, 5199 (1990)M.S. Stack, and R.D. Gray, J. Biol. Chem. 264, 4277 (1989)SKU(s): MMPS-020A, MMPS-020B, MMPS-020C, MMPS-020D
Catalog No. 50-210-9790 Supplier CPC Scientific Supplier No. MMPS020D
Efficient substrate for intestitial/vertebrate collagenase (MMP-1), stromelysin (MMP-3), gelatinases (MMP-2 and MMP-9) and the punctuated metalloproteinase (MMP-7).SEQUENCE: DNP-Pro-Leu-Gly-Leu-Trp-Ala-DArg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dnp-PLGLWA-DArg-NH2MOLECULAR FORMULA: C45H64N14O11MOLECULAR WEIGHT: 977.09STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [121282-17-5]SYNONYMS: Dnp-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg-NHRESEARCH AREA: CancerREFERENCES:D.M. Bickett, et al., Anal. Biochem. 212, 58 (1993)C.G. Knight, et al., FEBS Lett. 296, 263 (1992)K. Darlak, et al., J. Biol. Chem. 265, 5199 (1990)M.S. Stack, and R.D. Gray, J. Biol. Chem. 264, 4277 (1989)SKU(s): MMPS-020A, MMPS-020B, MMPS-020C, MMPS-020D