Learn More
CPC Scientific DABCYL-GABA-Pro-Gln-Gly-Leu-Glu(EDANS)-Ala-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109792
Encompass
Highly soluble fluorogenic matrix metalloproteinase (MMP) substrate with the following kcat/km values (at pH 7.6, 37C): 619.0 M-1s-1 for MMP-2, 209.0 M-1s-1 for MMP-9, 40.0 M-1s-1 for MMP-3, 21.0 M-1s-1 for MMP-1. The fluorescence increase is measured using an wavelength of 340 nm and emission wavelength of 485 nm. The relative high emission wavelength of EDANS allows convenient measurement of MMP activity in complex biological media like synovial fluid and culture medium.SEQUENCE: DABCYL-GABA-Pro-Gln-Gly-Leu-Glu(EDANS)-Ala-Lys-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dabcyl-GABA-PQGL-Glu(EDANS)-AK-NH2MOLECULAR FORMULA: C63H88N16O14S1MOLECULAR WEIGHT: 1325.56STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [193475-71-7]SYNONYMS: TNO211, DABCYL--Abu-Pro-Gln-Gly-Leu-Glu(EDANS)-Ala-Lys-NHRESEARCH AREA: CancerREFERENCES:B. Beekman, et al., FEBS Lett, 390, 221 (1996)SKU(
Catalog No. 50-210-9792 Supplier CPC Scientific Supplier No. MMPS021B
Highly soluble fluorogenic matrix metalloproteinase (MMP) substrate with the following kcat/km values (at pH 7.6, 37C): 619.0 M-1s-1 for MMP-2, 209.0 M-1s-1 for MMP-9, 40.0 M-1s-1 for MMP-3, 21.0 M-1s-1 for MMP-1. The fluorescence increase is measured using an wavelength of 340 nm and emission wavelength of 485 nm. The relative high emission wavelength of EDANS allows convenient measurement of MMP activity in complex biological media like synovial fluid and culture medium.SEQUENCE: DABCYL-GABA-Pro-Gln-Gly-Leu-Glu(EDANS)-Ala-Lys-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Dabcyl-GABA-PQGL-Glu(EDANS)-AK-NH2MOLECULAR FORMULA: C63H88N16O14S1MOLECULAR WEIGHT: 1325.56STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [193475-71-7]SYNONYMS: TNO211, DABCYL--Abu-Pro-Gln-Gly-Leu-Glu(EDANS)-Ala-Lys-NHRESEARCH AREA: CancerREFERENCES:B. Beekman, et al., FEBS Lett, 390, 221 (1996)SKU(