Learn More
CPC Scientific FITC-C6-Pro-Leu-Ala-Leu-Trp-Ala-Arg-Lys(Biotin)-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109795
Encompass
ONE-LETTER SEQUENCE: FITC-C6-PLALWAR-Lys(Biotin)-NH2MOLECULAR FORMULA: C83H112N18O16S2MOLECULAR WEIGHT:1682.02STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: E.J. Jacobsen, et al., J. Med. Chem. 42, 1525 (1999); J.V. Manetta, et al., Anal. Biochem. 202, 10 (1992)
Catalog No. 50-210-9795 Supplier CPC Scientific Supplier No. MMPS023A
ONE-LETTER SEQUENCE: FITC-C6-PLALWAR-Lys(Biotin)-NH2MOLECULAR FORMULA: C83H112N18O16S2MOLECULAR WEIGHT:1682.02STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: E.J. Jacobsen, et al., J. Med. Chem. 42, 1525 (1999); J.V. Manetta, et al., Anal. Biochem. 202, 10 (1992)