Learn More
Aviva Systems Biology Corp MMP1 Antibody - N-terminal region (ARP42039_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42039_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MMP1. Alternate names: CLG, CLGNApplicationIHC, WB. Species reactivity: Horse, Human
Catalog No. 50-014-58089