Learn More
ABclonal Technology MMP1 Rabbit mAb
SDP
Catalog No. 5000007592
Encompass_Preferred
This gene encodes a member of the peptidase M10 family of matrix metalloproteinases (MMPs) Proteins in this family are involved in the breakdown of extracellular matrix in normal physiological processes such as embryonic development reproduction and tissue remodeling as well as in disease processes such as arthritis and metastasis The encoded preproprotein is proteolytically processed to generate the mature protease This secreted protease breaks down the interstitial collagens including types I II and III The gene is part of a cluster of MMP genes on chromosome 11 Mutations in this gene are associated with chronic obstructive pulmonary disease (COPD) Alternative splicing results in multiple transcript variants at least one of which encodes an isoform that is proteolytically processed
Catalog No. 50-000-07592 Supplier ABclonal Technology Supplier No. A22080100UL
This gene encodes a member of the peptidase M10 family of matrix metalloproteinases (MMPs) Proteins in this family are involved in the breakdown of extracellular matrix in normal physiological processes such as embryonic development reproduction and tissue remodeling as well as in disease processes such as arthritis and metastasis The encoded preproprotein is proteolytically processed to generate the mature protease This secreted protease breaks down the interstitial collagens including types I II and III The gene is part of a cluster of MMP genes on chromosome 11 Mutations in this gene are associated with chronic obstructive pulmonary disease (COPD) Alternative splicing results in multiple transcript variants at least one of which encodes an isoform that is proteolytically processed