Learn More
Aviva Systems Biology Corp MMP13 Antibody - middle region (ARP56350_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56350_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MMP13. Alternate names: CLG3, MANDP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-61825