Learn More
Medchemexpress LLC HY-124029 5mg , MMP13-IN-3 CAS:1222173-37-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1240295MG
Encompass_Preferred
HY-124029 5mg MMP13-IN-3 CAS: 1222173-37-6 MMP13-IN-3 is a potent, selective, and orally active MMP-13 inhibitor (IC50=1 nM) for the potential treatment of osteoarthritis. MMP13-IN-3 is >1000 selective over other MMPs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1712