Learn More
Aviva Systems Biology Corp MMP16 Antibody - N-terminal region (ARP46674_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46674_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MMP16. Alternate names: MMP-X2, MT-MMP2, MT-MMP3, MT3-MMP, C8orf57ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59374