Learn More
Aviva Systems Biology Corp MMP19 Antibody - C-terminal region (ARP32315_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32315_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP19. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MMP19. Alternate names: MMP18, RASI-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54524