Learn More
Aviva Systems Biology Corp MMP19 Antibody - N-terminal region (ARP32314_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32314_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP19. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MMP19. Alternate names: MMP18, RASI-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54523