Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4313MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: MMP2/1501. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: U-138 MG or U-87 MG cells. Placenta. Cellular Staining: Cytoplasmic and Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human MMP2 protein fragment (aa444-575). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 72kDa (Pro) ;63kDa (cleaved).Chromosome: 16q12.2. Gene Symbol: MMP2. Gene ID: 4313. Ensemble: ENSG00000087245. Uniprot: NP_001289438.
Catalog No. 50-188-4942