Learn More
Aviva Systems Biology Corp MMP2 Antibody - C-terminal region (AVARP20016_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP20016_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MMP2. Alternate names: CLG4, MONA, CLG4A, TBE-1, MMP-IIApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-62650