Learn More
Cusabio Technology LLC MMP2/Gelatinase A
Catalog No. 5000051162
Encompass_Preferred
This kit detects MMP2/Gelatinase A in canine serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 4 69 ng/mL to 300 ng/mL with a sensitivity of 1 17 ng/mL ensuring precise measurement of target protein levels The assay requires 50-100 L of sample per test and employs a straightforward protocol with an incubation time of 1-5 hours Results are measured at 450 nm providing reliable data for research applications focused on gelatinase A activity in canine biological samples Designed for flexibility and accuracy this kit is suitable for researchers analyzing MMP2-related pathways in disease models or biomarker studies
Catalog No. 50-000-51162 Supplier Cusabio Technology LLC Supplier No. CSBE14056C
This kit detects MMP2/Gelatinase A in canine serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 4 69 ng/mL to 300 ng/mL with a sensitivity of 1 17 ng/mL ensuring precise measurement of target protein levels The assay requires 50-100 L of sample per test and employs a straightforward protocol with an incubation time of 1-5 hours Results are measured at 450 nm providing reliable data for research applications focused on gelatinase A activity in canine biological samples Designed for flexibility and accuracy this kit is suitable for researchers analyzing MMP2-related pathways in disease models or biomarker studies