Learn More
Aviva Systems Biology Corp MMP23B Antibody - middle region (ARP46807_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46807_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP23B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MMP23B. Alternate names: MIFR, MIFR-1, MMP22, MMP23AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-59406