Learn More
Aviva Systems Biology Corp MMP26 Antibody - C-terminal region (ARP57558_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57558_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP26. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MMP26. Alternate names: MGC126590, MGC126592ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-286-331