Learn More
Aviva Systems Biology Corp MMP3 Antibody - middle region (ARP42041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MMP3. Alternate names: CHDS6, MGC126102, MGC126103, MGC126104, MMP-3, SL-1, STMY, STMY1, STR1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-973