Learn More
Sino Biological MMP-3 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10467MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MMP-3 (rh MMP-3; Catalog#10467-HNAE; AAA36321.1; Tyr 18-Thr 272). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7412