Learn More
Enzo Life Sciences Matrix metalloproteinase-3 (MMP-3) fluorometric drug discovery kit, GREEN (1x96wells)
SDP
Catalog No. 502010255
Encompass_Preferred
The MMP-3 Fluorometric (also known as fluorimetric)Drug Discovery Kit, GREEN is a complete assay system designed to screen MMP-3 inhibitors using a quenched fluorogenic MMP-3 substrate: 5-FAM-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(TQ2W)-NH2 [5-FAM=5-carboxyfluorescein; Nva=norvaline; TQ2W=quencher]. FAM fluorescence is thoroughly quenched by the TQ2W group until cleavage by MMPs separates the two moieties. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-3, a potential therapeutic target. The compound NNGH is also included as a prototypic control inhibitor. Alternative name: Stromelysin 1, Transin 1. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0255 Supplier Enzo Life Sciences Supplier No. BMLAK3030001
The MMP-3 Fluorometric (also known as fluorimetric)Drug Discovery Kit, GREEN is a complete assay system designed to screen MMP-3 inhibitors using a quenched fluorogenic MMP-3 substrate: 5-FAM-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(TQ2W)-NH2 [5-FAM=5-carboxyfluorescein; Nva=norvaline; TQ2W=quencher]. FAM fluorescence is thoroughly quenched by the TQ2W group until cleavage by MMPs separates the two moieties. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-3, a potential therapeutic target. The compound NNGH is also included as a prototypic control inhibitor. Alternative name: Stromelysin 1, Transin 1. Applications: Fluorescent detection, HTS Activity assay.